Packaging Bags with Closure
Supplier: BURKLE
2348-0006 2348-0008 2348-0010 2348-0003 2348-0001 2348-0004 2348-0007 2348-0002 2348-0009
BURK2348-0006EA 53.4 SGD
BURK2348-0006 BURK2348-0008 BURK2348-0010 BURK2348-0003 BURK2348-0001 BURK2348-0004 BURK2348-0007 BURK2348-0002 BURK2348-0009
Packaging Bags with Closure
Bags Storage Bags
Bags with the patented zipper are for ideal for storing, collecting, protecting and shipping of specimens, samples and sales products.
  • Made of LDPE
  • Film has good tear resistance
  • Crystal-clear
  • Waterproof
Contents remain dust-free.
Order Now
SPECIFICATIONS
Environmentally Preferable