Eyewash station for tap, EyesFresh
Supplier: BURKLE
2515-0100
BURK2515-0100EA 561 SGD
BURK2515-0100
Eyewash station for tap, EyesFresh
First Aid Supplies Eyewash Stations and Eyewash Bottles
This mini eyewash station is ideal for laboratories, filling stations or all work places where aggressive substances are used.
  • Flexible usage as it can be attached to most water taps
  • No refilling needed as the water from the tap is used
It can be easily attached to any standard cold water tap. For normal use of the tap, the user just needs to press the button and the function changes back from the eye wash (returns automatically to eyewash-function when the tap is closed).
Can be fitted onto taps with M22×1, 1/2" or 3/8" outer thread.