Samplers, PowderProof®
Supplier: BURKLE
5302-0302 5302-0602 5302-0902 5302-1202 5302-0301 5302-0601 5302-0901 5302-1201 5302-0316 5302-0317 5302-0617 5302-0616 5302-0917 5302-0916 5302-1217 5302-1216
BURK5302-0302EA 8440 SGD
BURK5302-0302 BURK5302-0602 BURK5302-0902 BURK5302-1202 BURK5302-0301 BURK5302-0601 BURK5302-0901 BURK5302-1201 BURK5302-0316 BURK5302-0317 BURK5302-0617 BURK5302-0616 BURK5302-0917 BURK5302-0916 BURK5302-1217 BURK5302-1216
Samplers, PowderProof®
Samplers Solid Samplers
Quality assurance starts with the inspection of incoming goods. An all-layer sample of bulk goods is no longer a problem with PowderProof®, the sampler for powders and granules.
  • Many uses
  • Stainless steel AISI 316L (1.4404)/AISI 301 (1.4310)
  • Includes motor, two rechargeable batteries, fast charger, lance, spiral screw, bags, bag clip, and cleaning brush
  • Additional spiral screws available as accessory
PowderProof® is the practically proven professional solution for mobile sampling of powders and granules. Different lances and augers can be attached toMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable