Labels, control seals, close-it
Supplier: BURKLE
5303-0011 5303-0012 5303-0014 5303-0016 5303-0002 5303-1012 5303-1011 5303-1014 5303-1015 5303-1016 5303-1002 5303-0015
BURK5303-0011EA 196 SGD
BURK5303-0011 BURK5303-0012 BURK5303-0014 BURK5303-0016 BURK5303-0002 BURK5303-1012 BURK5303-1011 BURK5303-1014 BURK5303-1015 BURK5303-1016 BURK5303-1002 BURK5303-0015
Labels, control seals, close-it
Labels
Designed to seal the holes in sacks or boxes created when samples are taken, the strong adhesive on the 'close-it' label allows adhesion to surfaces that are slightly moist, frozen or coated with powder. The aluminium backing creates a vapour barrier and seals the container hermetically.
  • Adheres extremely firmly
  • Can be written on
  • Colour coded for easy recognition
Delivery information: Rolls contain 500 (95×95 mm) or 250 (150×150 mm) stickers.