Control seal, close-it tape
Supplier: BURKLE
5303-3019 5303-2019 5303-0017 5303-1017
BURK5303-3019EA 286 SGD
BURK5303-3019 BURK5303-2019 BURK5303-0017 BURK5303-1017
Control seal, close-it tape
Labels
Standard close-it and close-it food & pharma control seals are also available on a continuous roll. This offers the flexibility to provide a small length of tape to seal a small hole or to mark a specific point. Alternatively, longer pieces can be used to repair a long tear quickly, reliably and easily. With its extremely strong adhesive strength, close-it tapes adhere to a range of different surfaces, where the glue effect of normal adhesive tape quickly reaches its maximum capacity.
  • Customisable
  • White with tear-off lines
  • Available in the classic
More Product Information