Control seals, close-it food & pharma
Supplier: BURKLE
5303-2017 5303-2018 5303-3018 5303-3017 5303-2020 5303-2023 5303-2021 5303-2022 5303-3020 5303-3022 5303-3021 5303-3023 9990-8075
BURK5303-2017EA 196 SGD
BURK5303-2017 BURK5303-2018 BURK5303-3018 BURK5303-3017 BURK5303-2020 BURK5303-2023 BURK5303-2021 BURK5303-2022 BURK5303-3020 BURK5303-3022 BURK5303-3021 BURK5303-3023 BURK9990-8075
Control seals, close-it food & pharma
Labels
A control seal with an adhesive approved for use with foodstuffs. The strength of the adhesive is almost as powerful as that of the original. Just like the original, close-it food and pharma has a barrier layer of aluminium foil.
  • Adheres very firmly
  • Can be written on
  • Adhesive complies with the FDA and European guidelines for foodstuffs
Suitable for sampling foodstuffs, pharmaceuticals, cosmetics, food additives, even frozen goods where cleanliness is particularly important.
Order Now
SPECIFICATIONS
Environmentally Preferable