Tamper proof labels, safe-it
Supplier: BURKLE
5303-5000
BURK5303-5000EA 324 SGD
BURK5303-5000
Tamper proof labels, safe-it
Labels
These safety labels are used as originality seals and as protection from manipulations of e.g. samples, retention samples, for transporting samples, sealing containers with important or dangerous chemicals in the lab as well as sealing letters, shipments and much more. The initial opening of a package is made obvious and irrevocably visible with the seal guaranteeing originality, unauthorised opening can therefore be proven.
  • With checkerboard pattern
  • Visible proof of tampering
If the safety label is removed, it divides into separate layers. A patternMore Product Information