Powder samplers, MicroSampler
Supplier: BURKLE
5307-2055 5307-0205 5307-2085 5307-0210 5307-1055 5307-1085 5307-1120 5307-0102 5307-0105 5307-0110 5307-2120 5307-0202 5307-0101
BURK5307-2055EA 1170 SGD
BURK5307-2055 BURK5307-0205 BURK5307-2085 BURK5307-0210 BURK5307-1055 BURK5307-1085 BURK5307-1120 BURK5307-0102 BURK5307-0105 BURK5307-0110 BURK5307-2120 BURK5307-0202 BURK5307-0101
Powder samplers, MicroSampler
Samplers Solid Samplers
Stainless steel AISI 316 L (1,4404). The MicroSampler has been specially designed for the sampling of small quantities of powder.
  • Point sampling
  • Various lengths and volumes are available
  • Tip can be removed for easy cleaning
  • Additional tips are available in different sizes
  • Sterilisable
Volumes of 0,2 ml, 0,5 ml, 1,0 ml, 2,0 ml, 5,0 ml, and 10 ml can be sampled easily, quickly, and cleanly from different sampling depths from sacks or containers.
The tips of the MicroSampler are interchangeable. This means that tips with different volumes can be used.
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable