Sampler with suction lance, UniSampler
Supplier: BURKLE
5314-1000
BURK5314-1000EA 795 SGD
BURK5314-1000
Sampler with suction lance, UniSampler
Samplers Fluid Samplers
The collection lance consists of an inner tube and an outer tube. The suction position can be varied steplessly with the outer tube within a height of 0 to 48 cm.
  • PP suction tube and PVC suction hose
  • Universal bottle adapter made of PVC
  • Bottle necks Ø 24, 32, 39 and 50 mm nominal dimension
Delivery information: Supplied with five PP sample bottles of 250 ml and 500 ml, a vacuum pump, an adapter for sample bottles 100 to 1000 ml, 20 seal-it bottle seals.