Sampler, UniSampler 'Ex'
Supplier: BURKLE
5314-3000
BURK5314-3000EA 1300 SGD
BURK5314-3000
Sampler, UniSampler 'Ex'
Samplers Fluid Samplers
Glass bottle and hose are connected in such a way that they conduct electricity and are grounded via a grounding cable. Operation: Ground the device, immerse the suction hose to the required depth, attach the adapter to the sampling bottle, generate a vacuum with the manual pump and collect the sample, remove the filled sampling bottle, close it and label it.
  • Specially developed for sampling flammable liquids of hazardous materials (e.g. gasoline)
  • PTB approval D- 333-009 618/00
Specially developped for sampling flammable liquids of hazardousMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable