Sample dippers
Supplier: BURKLE
5315-0050 5315-0200 5315-0100 5315-0300
BURK5315-0050EA 158 SGD
BURK5315-0050 BURK5315-0200 BURK5315-0100 BURK5315-0300
Sample dippers
Samplers Sample Dippers
Plunging siphons for the simple removal of liquid samples.
  • Made from transparent PP
  • All-layer and point sampling
  • Upper end with reflux valve, lower end with ball valve
Order Now
SPECIFICATIONS
Environmentally Preferable