Sample spoons
Supplier: BURKLE
5320-0150 5320-0180 5320-0210 5320-0300 5320-0400 5320-0500
BURK5320-0150EA 13.6 SGD
BURK5320-0150 BURK5320-0180 BURK5320-0210 BURK5320-0300 BURK5320-0400 BURK5320-0500
Sample spoons
Spoons
Made of AISI 304 (1.4301) stainless steel. This specially shaped spoon reaches almost every corner in containers and prevents samples from trickling out.
  • For withdrawing small sample quantities
  • Largely chemical resistant (not against acids and alkalis)