Spoons, PharmaSpoon
Supplier: BURKLE
5324-0001
BURK5324-0001EA 1870 SGD
BURK5324-0001
Spoons, PharmaSpoon
Spoons
Stainless steel V4A (1.4404), high polish finish. For taking samples of powdery substances, intermediate products and material in the pharmaceutical industry.
  • Possible residues are even visible in the trace sector (µg sector)
  • Also suitable for sterile sampling
  • Made of materials with excellent surface qualities to prevent cross-contamination
Order Now
SPECIFICATIONS
Environmentally Preferable