Samplers, Liquid-Sampler
Supplier: BURKLE
5326-0001 5326-0002 5326-0004 5326-0003
BURK5326-0001EA 758 SGD
BURK5326-0001 BURK5326-0002 BURK5326-0004 BURK5326-0003
Samplers, Liquid-Sampler
Samplers Fluid Samplers
Material: V4A stainless steel (1.4404). Sampler for thin to viscous liquids.
  • Easy one handed operation
  • Discharge funnel for safe filling of sample bottles