Sampler, Vampire
Supplier: BURKLE
5327-1000
BURK5327-1000EA 4140 SGD
BURK5327-1000
Sampler, Vampire
Samplers Fluid Samplers
The Vampire sampler can be used to take samples from liquids easily, even from deeper depths.
  • Battery-powered hose pump for sampling liquids from depth
  • Easy cleaning and exchangeable hoses prevent impurities
  • Suction depth up to 5 meters
  • Portable sampler is easy to handle and carry
  • Pump attachment is easy to attach to the drive unit
  • Sample bottle is fastened directly onto the sampling instrument by a bottle basket
The portable pump head consists of a mains-independent drive unit, a pump head, and a delivery hose.
The medium being sampled is directedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable