Fluid samplers, ViscoSampler
Supplier: BURKLE
5331-2100 5331-2200 5331-1100 5331-1200 5331-1060 5331-2060
BURK5331-2100EA 1340 SGD
BURK5331-2100 BURK5331-2200 BURK5331-1100 BURK5331-1200 BURK5331-1060 BURK5331-2060
Fluid samplers, ViscoSampler
Samplers Fluid Samplers
ViscoSampler with powerful suction for viscous media. The media is drawn up and discharged using the suction flask.
  • Made of ultra-pure, chemically inert PTFE/FEP for contamination-free sampling
  • Easy cleaning - all surfaces are pore less and without crevices
  • Dirt accumulation is prevented
  • Only round screw threads tried and tested in the field of food hygiene are used
  • Disassembly and cleaning are extremely simple
Specially developed for viscous substances such as slurries, silt, wet clay and soil samples, soil sediments, oils, emulsions and creams.
Order Now
SPECIFICATIONS
Environmentally Preferable