Sampler, SiloPicker
Supplier: BURKLE
5340-1100
BURK5340-1100EA 1450 SGD
BURK5340-1100
Sampler, SiloPicker
Samplers Solid Samplers
For sampling bulk goods at depths of up to 3,5 metres, depending on the density of the bulk goods to be sampled. Inserting the sampler into the bulk goods closes the collection chamber. At the desired depth, the collection chamber is opened by withdrawing the sampler, the chamber is filled and can be pulled out and collected.
  • Easy handling
  • Sampler length 1300 mm, may be extended up to 3500 mm using extension rods
  • Stainless steel V4A (1.4404)/AISI 316 L
Order Now
SPECIFICATIONS
Environmentally Preferable