Sample bags with closure, SteriBag
Supplier: BURKLE
5344-0540 5344-0720 5344-0180 5344-0721 5344-0060 5344-0810 5344-1650 5344-0061 5344-0181 5344-0541 5344-0811 5344-1651
BURK5344-0540EA 596 SGD
BURK5344-0540 BURK5344-0720 BURK5344-0180 BURK5344-0721 BURK5344-0060 BURK5344-0810 BURK5344-1650 BURK5344-0061 BURK5344-0181 BURK5344-0541 BURK5344-0811 BURK5344-1651
Sample bags with closure, SteriBag
Bags Sample Bags
The sterilised sampling bag SteriBag are ideal for samples in chemistry, pharmaceuticals, medicine, veterinary science and foodstuffs.
  • Tearproof and leakproof
  • Can be labelled
Tab-ends are sealed in plastic film to prevent skin injuries.
Order Now
SPECIFICATIONS
Environmentally Preferable