Scratcher for sampling
Supplier: BURKLE
5347-0010 5347-0005 5347-0025
BURK5347-0010EA 4760 SGD
BURK5347-0010 BURK5347-0005 BURK5347-0025
Scratcher for sampling
Samplers Solid Samplers
The scratcher has particularly been designed for sampling cohesive or caked substances.
  • Electro-polished stainless steel
For non pourable substances.