Soil samplers, GeoSampler
Supplier: BURKLE
5350-5003 5350-5006
BURK5350-5003EA 426 SGD
BURK5350-5003 BURK5350-5006
Soil samplers, GeoSampler
Samplers Solid Samplers
GeoSampler is specially designed for soil sampling.
  • Depth graduation 10 cm divisions
  • With or without foot bar for easier work
Sampler for light soils, meadows and lawns.
Order Now
SPECIFICATIONS
Environmentally Preferable