Soil samplers, Puerkhauer
Supplier: BURKLE
5350-5012 5350-5018
BURK5350-5012EA 1450 SGD
BURK5350-5012 BURK5350-5018
Soil samplers, Puerkhauer
Samplers Solid Samplers
Very robust sampler for heavy and stony soils. The model with 1000 mm flute length has a conically-tapered 60 cm tip which makes it much easier to extract the driven-in sampler.
  • Depth graduation of 10 cm divisions
  • Attachable handle and non-slip plastic grips
  • Reinforced impact head