Scoop
Supplier: BURKLE
5354-1000
BURK5354-1000EA 1060 SGD
BURK5354-1000
Scoop
Scoops
Stainless steel V2A (1.4301).
  • Complete with rod
  • No openings, undercuts or crevices in which residues could collect
  • Pore-free finish produced by high quality laser welding seams
  • Sterilisable by flame or steam sterilisation
Order Now
SPECIFICATIONS
Environmentally Preferable