Samplers for liquids, TeleScoop
Supplier: BURKLE
5354-0100 5354-0200 5355-0450 5354-4100 5354-0050 5354-5050 5354-5100 5354-5200 5355-0250 5354-2000 5354-3000 5355-0120 5355-0280 5355-0300 5355-0600 5354-5010 5354-5020 5355-0180 5354-2010
BURK5354-0100EA 162 SGD
BURK5354-0100 BURK5354-0200 BURK5355-0450 BURK5354-4100 BURK5354-0050 BURK5354-5050 BURK5354-5100 BURK5354-5200 BURK5355-0250 BURK5354-2000 BURK5354-3000 BURK5355-0120 BURK5355-0280 BURK5355-0300 BURK5355-0600 BURK5354-5010 BURK5354-5020 BURK5355-0180 BURK5354-2010
Samplers for liquids, TeleScoop
Samplers Fluid Samplers
TeleScoop is a versatile sampling system with exchangeable tools for a wide range of applications. The tools (angular beaker, pendulum beaker, bottle holder, stainless steel beaker and dip net) are attached to the telescopic rod by a practical snap-in joint. Ideal for sampling from baths, tanks, shafts, open water courses. Sampling depths down to 6 m can be reached with the adjustable telescopic aluminium rod.
  • For industry and water treatment
Angular beaker adaptable to different container shapes by adjustable angle setting (up to 90°).
PP pendulumMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable