Dipping bottle, 'Ex'
Supplier: BURKLE
5365-3000
BURK5365-3000EA 1870 SGD
BURK5365-3000
Dipping bottle, 'Ex'
Samplers Sample Dippers
Dipping bottle for testing mineral oil products (Category A, Explosion Categories IIA, IIB and IIC), crude oil, liquid mineral oil products, fuels, special and test fuels, fuel oils, petroleum and liquid lubricants. Completely made from non spark-producing materials, nickel plated.
  • Screw-off bottle head
  • Easy to clean robust handle
  • High weight (3,2 kg) for fast lowering
Certifications: Tested for use in the Ex protection zone 0.
Ordering information: Please order the appropriate lowering cable at the same time.
Order Now
SPECIFICATIONS
Environmentally Preferable