Mini immersion cylinder
Supplier: BURKLE
5365-5000
BURK5365-5000EA 1120 SGD
BURK5365-5000
Mini immersion cylinder
Samplers Fluid Samplers
Mini immersion cylinder for sampling liquids in narrow places where access is difficult.
  • Screw-off top enables easy cleaning
  • Tested for use in the Ex protection zone 0
  • Explosion Categories IIA and IIB
Can also be used in curved dipping tubes and barrels with narrow openings.
Ordering information: Please order the appropriate lowering cable at the same time.