Sampling scoops, SteriPlast®
Supplier: BURKLE
5378-1007 5378-1001 5378-1002 5378-1003 5378-1004 5378-1005 5378-1006 5378-1008 5378-1013 5378-1014 5378-1015 5378-1016 5378-1019 5378-1020
BURK5378-1007EA 66.5 SGD
BURK5378-1007 BURK5378-1001 BURK5378-1002 BURK5378-1003 BURK5378-1004 BURK5378-1005 BURK5378-1006 BURK5378-1008 BURK5378-1013 BURK5378-1014 BURK5378-1015 BURK5378-1016 BURK5378-1019 BURK5378-1020
Sampling scoops, SteriPlast®
Scoops
PS, white. These scoops are ideal for the sampling of powder, granules and pastes. They are also available with a transparent lid to close it immediately after the sampling procedure to secure a sterile and contamination-free sample.
  • Individually packed, gamma radiation sterilised
  • Production, assembling and packaging acc. to ISO cleanroom class 7
  • According to EU food and FDA guidelines
Due to the special handle construction and the slightly angled edge the scoop stands horizontal and can be laid down without the material trickling out.
Order Now
SPECIFICATIONS
Environmentally Preferable