Spoon spatulas, LaboPlast® / SteriPlast®
Supplier: BURKLE
5378-0031 5378-1031
BURK5378-0031EA 71.1 SGD
BURK5378-0031 BURK5378-1031
Spoon spatulas, LaboPlast® / SteriPlast®
Spatulas
PS. Double spatula with a 0,5 ml spoon and a 17 mm spatula blade. Ideal for use as a sampling spoon, for measuring or transferring small amounts, as a scraper or for mixing and stirring.
  • Resistant to chemicals, corrosion-proof
  • Production, assembly and packaging according to cleanroom class 7
  • Complies with EU food requirements and FDA regulations
  • Optionally packaged individually and sterilised by gamma rays
Packaging: Available either in a large package with 100 pieces or 100 pieces packaged individually and sterilised by gamma rays.
Order Now
SPECIFICATIONS
Environmentally Preferable