Detectable scoops, SteriPlast®
Supplier: BURKLE
5378-2001 5378-2003 5378-2005 5378-2007 5378-2013 5378-2015 5378-2019 5378-2020
BURK5378-2001EA 87.1 SGD
BURK5378-2001 BURK5378-2003 BURK5378-2005 BURK5378-2007 BURK5378-2013 BURK5378-2015 BURK5378-2019 BURK5378-2020
Detectable scoops, SteriPlast®
Scoops
These detectable SteriPlast® scoops, made of blue PS, are produced and assembled in a class 7 cleanroom (10,000), individually packaged for disposable use and then sterilised by gamma rays. Detectable scoops are ideal for powders, granulates and pastes.
  • Can be detected by colour scanners, metal detectors or X-rays
  • Individually packaged and sterilised by gamma rays
  • Production, assembly and packaging according to cleanroom class 7
  • Complies with EU food requirements and FDA regulations
These scoops are not only coloured blue but also have a specialMore Product Information