Detectable spatulas, SteriPlast®
Supplier: BURKLE
5378-2009
BURK5378-2009EA 81.7 SGD
BURK5378-2009
Detectable spatulas, SteriPlast®
Spatulas
These detectable SteriPlast® spatulas, made of blue PS, are produced and assembled in a class 7 cleanroom (10,000), individually packaged for disposable use and then sterilised by gamma rays. Ideal for sampling powders, granulates and pastes.
  • Can be detected by colour scanners, metal detectors or X-rays
  • Individually packaged and sterilised by gamma rays
  • Production, assembly and packaging according to cleanroom class 7
  • Complies with EU food requirements and FDA regulations
These spatulas are not only coloured blue but also have a special additive inMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable