Spatulas for foodstuffs, SteriPlast®
Supplier: BURKLE
5378-3009
BURK5378-3009EA 53.9 SGD
BURK5378-3009
Spatulas for foodstuffs, SteriPlast®
Spatulas
The SteriPlast® spatula for foodstuffs, made of PS, is produced and assembled in a class 7 cleanroom (10,000), individually packaged for disposable use and then sterilised by gamma rays. The spatula is ideal for sampling powders, granulates and pastes.
  • Can be used as part of HACCP/IFS foreign object management
  • Especially for the food, animal feed and pharmaceutical industries
  • Individually packaged and sterilised by gamma rays
  • Production, assembly and packaging according to cleanroom class 7
  • Complies with EU food requirements and FDA regulations
Order Now
SPECIFICATIONS
Environmentally Preferable