Spoons for foodstuffs, SteriPlast®
Supplier: BURKLE
5378-3011 5378-3012
BURK5378-3011EA 138 SGD
BURK5378-3011 BURK5378-3012
Spoons for foodstuffs, SteriPlast®
Spoons
The SteriPlast® spoon for foodstuffs, made of PS, is produced in a class 7 cleanroom (10,000), individually packaged for disposable use and then sterilised by gamma rays. The spoon is available in two sizes, 2,5 ml (similar to a teaspoon) and 10 ml (similar to a tablespoon).
  • Can be used as part of HACCP/IFS foreign object management
  • Especially for the food, animal feed and pharmaceutical industries
  • Individually packaged and sterilised by gamma rays
  • Production, assembly and packaging according to cleanroom class 7
  • Complies with EU food requirements and FDA
More Product Information