Dosing spoons, white
Supplier: BURKLE
5378-4014 5378-4015 5378-4017 5378-4019 5378-4006 5378-4007 5378-4011 5378-4012 5378-4013 5378-4010 5378-4016 5378-4018 5378-4004 5378-4008 5378-4009 5378-4005
BURK5378-4014EA 109 SGD
BURK5378-4014 BURK5378-4015 BURK5378-4017 BURK5378-4019 BURK5378-4006 BURK5378-4007 BURK5378-4011 BURK5378-4012 BURK5378-4013 BURK5378-4010 BURK5378-4016 BURK5378-4018 BURK5378-4004 BURK5378-4008 BURK5378-4009 BURK5378-4005
Dosing spoons, white
Spoons
Volumetric and dosing spoons can be used to measure small amounts of powder, granulates and liquids exactly and dose them precisely. For precise measuring, excess medium is stripped off the upper edge of the spoon. The content volume is engraved on the handle.
  • PS, white
  • Optionally packed individually and sterilised
  • Stripper edge
  • For single use
  • Cleanroom manufactured (cleanroom Class 7)
The flat front edge of the volumetric spoon is ideal for picking up medium from the bottom or getting into the corners of a container, thus allowing for efficientMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable