Dosing spoon and measuring spoon set
Supplier: BURKLE
5378-4125 5378-4120
BURK5378-4125EA 29.7 SGD
BURK5378-4125 BURK5378-4120
Dosing spoon and measuring spoon set
Spoons
Dosing and measuring spoons can be used to measure small amounts of powder, granulates and liquids exactly and dose them precisely. For precise measuring, excess medium is stripped off the upper edge of the spoon. The content volume is engraved on the handle.
  • Content volume: 0,5; 1,0; 2,5; 5,0; 10; 15; 25; and 50 ml
  • Set of volumetric spoons with eight sizes
  • PS
  • With stripper edge
The flat front edge of the volumetric spoon is ideal for picking up medium from the bottom of the container or for getting into the corners of the container, thus allowing forMore Product Information