Food spoons, long handle, blue, SteriPlast®
Supplier: BURKLE
5378-5003 5378-5007
BURK5378-5003EA 66.5 SGD
BURK5378-5003 BURK5378-5007
Food spoons, long handle, blue, SteriPlast®
Spoons
The sampling spoon with long handle can be used to remove powders, granulates, pastes or liquids. The long handle makes it easy to reach the bottoms of larger containers as well and allows for sampling of drums, jerrycans and bottles with small opening diameters. Thanks to the indentations on the handle, the disposable spoon rests comfortably and securely in the hand.
  • PS, blue
  • Individually packaged and sterilised, for single use
  • Can be used as part of HACCP/IFS/BRC foreign object management
  • Especially for the food, animal feed and pharmaceutical industries
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable