Sampling spoons curved, long handle, LaboPlast®/SteriPlast®
Supplier: BURKLE
5378-5010 5378-5009
BURK5378-5010EA 69.9 SGD
BURK5378-5010 BURK5378-5009
Sampling spoons curved, long handle, LaboPlast®/SteriPlast®
Spoons
The spoon with curved spoon bowl is suitable for a wide range of different media including powders, granulates, pastes and liquids.
  • PS, white
  • Individually packaged and optionally sterilised
  • With curved spoon shape
  • Handle length 300 mm
  • For single use
  • Clean room manufactured (clean room class 7)
The angled spoon shape is also good for reaching into the corners of a container. The long handle of the plastic spoon makes it possible to empty residue from large containers easily and can also reach into places that are hard to reach such as jerrycans withMore Product Information