Sampling spoons, LaboPlast® Bio / SteriPlast® Bio
Supplier: BURKLE
5379-0011 5379-1011 5379-1012 5379-0012
BURK5379-0011EA 148 SGD
BURK5379-0011 BURK5379-1011 BURK5379-1012 BURK5379-0012
Sampling spoons, LaboPlast® Bio / SteriPlast® Bio
Spoons
Bio PE, white. These disposable spoons are manufactured from bio-plastic 'Green PE' which is entirely made of renewable resources.
  • 'Green PE' is 100% recyclable, made without fossil raw materials
  • Available non sterile (LaboPlast® Bio) or gamma radiation sterilised (SteriPlast® Bio)
  • Available in two sizes: 2,5 ml (similar to teaspoon) and 10 ml (similar to tablespoon)
  • Produced in a cleanroom environment
  • Individually packed
They are ideal for the sampling of powder, granules and pastes.