Labels, seal-it
Supplier: BURKLE
5399-0003
BURK5399-0003EA 464 SGD
BURK5399-0003
Labels, seal-it
Labels
Retention samples are kept to provide a sample for comparison at a later date. The unambiguous proof that the retention sample is identical to the original sample is only possible with clearly marked and sealed containers. However, sealing is frequently not possible unless eyelets are present. seal-it provides the solution! The seal-it self-adhesive label sticks to all conventional containers. Thanks to its unbelievable adhesive power it cannot be removed. If a container opening is closed with seal-it, then the container cannot be opened without destroying the seal-it label. seal-it isMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable