Safety pipetting-ball
Supplier: BURKLE
7554-0002
BURK7554-0002EA 60.8 SGD
BURK7554-0002
Safety pipetting-ball
Pipettes Pipette Bulbs
Peleus ball, simple pipetting aid. Evacuation takes place via an automatic valve. By pressing the valves, the liquid is drawn up, dispensed or purged.
  • For all pipettes up to 100 ml
The removable automatic valve also enables effective cleaning of the inside.