Tubing clamps, Squeeze-Fix
Supplier: BURKLE
8618-0002 8618-0003 8618-0001
BURK8618-0002EA 4.8 SGD
BURK8618-0002 BURK8618-0003 BURK8618-0001
Tubing clamps, Squeeze-Fix
Clamps Tubing and Sanitary Clamps
Permanently elastic hose clamps made of white POM.
  • Hose can be squeezed in phases or completely compressed, as required
  • Can be operated with one hand
  • Sterilisable, do not rust