Tubing clamps, PVDF, LaboPlast®
Supplier: BURKLE
8619-0104 8619-0102 8619-0202 8619-0206 8619-0154 8619-0155 8619-0105 8619-0106 8619-0152 8619-0156 8619-0205 8619-0204
BURK8619-0104EA 80.2 SGD
BURK8619-0104 BURK8619-0102 BURK8619-0202 BURK8619-0206 BURK8619-0154 BURK8619-0155 BURK8619-0105 BURK8619-0106 BURK8619-0152 BURK8619-0156 BURK8619-0205 BURK8619-0204
Tubing clamps, PVDF, LaboPlast®
Clamps Tubing and Sanitary Clamps
PVDF, aluminium and galvanised steel hook. The micro adjustment screw enables the flow of liquids to be controlled delicately and smoothly or stopped completely. The clamp can also be mounted on existing tube connections. The clamp's closure can be opened in any position, without screwing on and off.
  • Heat resistant up to +140 °C
  • Various sizes available for tubing from 1 to 20 mm Ext.Ø
  • Available in four colours