Non return valves
Supplier: BURKLE
8714-0100 8714-0060 8714-0080 8714-1008 8714-1006 8714-1010
BURK8714-0100EA 9.3 SGD
BURK8714-0100 BURK8714-0060 BURK8714-0080 BURK8714-1008 BURK8714-1006 BURK8714-1010
Non return valves
Valves Non Return Valves
Translucent PP retaining valves with embossed directional arrow or printed in black. With tapered ends. Various uses.
  • Resistant to compression max. 4 bar
  • Operating temperature up to +40 °C
  • With PP ball valve
Order Now
SPECIFICATIONS
Environmentally Preferable