Silicone rubber stoppers, conical
Supplier: BURKLE
9764-1115 9764-1621 9764-1924 9764-2227 9764-2834
BURK9764-1115EA 2.1 SGD
BURK9764-1115 BURK9764-1621 BURK9764-1924 BURK9764-2227 BURK9764-2834
Silicone rubber stoppers, conical
Stoppers Silicone Stoppers
These conical silicone rubber stoppers are translucent, chemically-biologically neutral, food-safe according to BfR Recommendation XV and FDA § 177.2600.
  • Temperature resistant from –60 to +200 °C, briefly to +230 °C
  • Sterilisable
  • Non-toxic
Order Now
SPECIFICATIONS
Environmentally Preferable