Tubular Bag Film for Sterilisation
Supplier: SÜDPACK MEDICA
7600519 7600523
CDPA7600519EA 380 SGD
CDPA7600519 CDPA7600523
Tubular Bag Film for Sterilisation
Bags Sterilisation Pouches
Tubular bag film is available as a flat or gussetted bag in various sizes. Helpful tips for use are printed on the film. Suitable for autoclaving and ethylene oxide sterilisation.
* Gusset's dimension.
Order Now
SPECIFICATIONS
Environmentally Preferable