Wound cleansers
Supplier: ORKLA CARE
3227
CEDE3227EA 28.1 SGD
CEDE3227
Wound cleansers
First Aid Supplies First Aid Kit Components
Wound cleansers do away with cotton wool and bottles of antiseptic. These sterile, non woven tissues, which are saturated in a wound cleansing fluid (0,9% NaCl), are intended for the mechanical cleansing of minor wounds when clean tap water is not available.
  • Tissues are packed in sealed sachets, which prevent leakage and guarantee sterility
  • Savett sachets are packed in a refill box
  • Salvequick cleansers fit into first-aid-stations