Adhesive plasters, Salvequick
Supplier: ORKLA CARE
6036 6444
CEDE6036EA 76.3 SGD
CEDE6036 CEDE6444
Adhesive plasters, Salvequick
First Aid Supplies Wound Dressings
These plaster refills are available in different versions: Made from plastic or fabric, blue detectable and for sensitive skin.
  • In combination with dispenser: Complete, easy to use system
  • Plasters are always available, ready to use
  • Plasters are allergy-tested
  • Each adhesive bandage is individually wrapped and sterile
Detectable plasters are specially designed for use in the food industry. The plasters are detectable by most metal detectors used within the food industry. The blue colour also makes the plasters easy to see.
Sensitive plasters areMore Product Information