Adapters, glass, flushing, Airfree, Schlenk type
Supplier: CHEMGLASS
AF-0509-11
CGLAAF-0509-11EA 397 SGD
CGLAAF-0509-11
Adapters, glass, flushing, Airfree, Schlenk type
Adapters Adapters, Conical Ground Joints
Adapter allows vessels to be flushed with inert gas while sampling directly through the bore of the T-Bore stopcock. With a 4 mm bore high vacuum glass stopcock with stainless steel constant pressure retention device.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Both the glass and PTFE stopcock have 9 mm OD medium wall arms for use with septum stopper.
Order Now
SPECIFICATIONS
Environmentally Preferable