Adapters, reduction
Supplier: CHEMGLASS
CG-1000-44
CGLACG-1000-44EA 131 SGD
CGLACG-1000-44
Adapters, reduction
Adapters Adapters, Conical Ground Joints
Used to connect dissimilar standard taper joint sizes. Reduction adapter from large cone at the base to small socket on the top.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes