Adapters, inlet, with cone
Supplier: CHEMGLASS
CG-1014-01
CGLACG-1014-01EA 95.3 SGD
CGLACG-1014-01
Adapters, inlet, with cone
Adapters Adapters, Joints with Tube Connectors
Adapters allow for flexible tubing to be connected to apparatus for introduction of vacuum or pressure. Hose connection has a 10 mm O.D. at the largest serration and is bent downward at a 90° angle to the cone.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Order Now
SPECIFICATIONS
Environmentally Preferable