Receiver adapters, vacuum take-off, bent
Supplier: CHEMGLASS
CG-1050-01
CGLACG-1050-01EA 214 SGD
CGLACG-1050-01
Receiver adapters, vacuum take-off, bent
Distillation Components Distillation Adapters
With standard ground joint cone and socket with sealed in drip tip. Adapter is bent at a 105° angle for use in vacuum distillation. Serrated hose connection has an O.D. of 10 mm at the largest serration for connection to vacuum source.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Order Now
SPECIFICATIONS
Environmentally Preferable