Beaker, low form, quartz
Supplier: CHEMGLASS
CG-1110-03
CGLACG-1110-03EA 588 SGD
CGLACG-1110-03
Beaker, low form, quartz
Beakers
Low form beaker made of quartz.
  • Non-graduated with pouring spout
  • Beaker can be used at temperatures to 1000 °C
  • Very good chemical resistance